Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILF2 Recombinant Rabbit Monoclonal Antibody (5P1K2)
GREENER_CHOICE

Catalog No. PIMA542922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542922 Supplier Invitrogen™ Supplier No. MA542922
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, HepG2, Raji, Mouse thymus, Rat testis. Immunogen sequence: SALAAIRHAR WFEENASQST VKVLIRLLKD LRIRFPGFEP LTPWILDLLG HYAVMNNPTR QPLALNVAYR RCLQILAAGL FLPGSVGITD PCESGNFRVH T.

Nuclear factor of activated T-cells is a transcription factor required for T-cell expression of the interleukin 2 gene. NFAT binds to a sequence in the interleukin 2 gene enhancer known as the antigen receptor response element 2. In addition, NFAT can bind RNA and is an essential component for encapsidation and protein priming of hepatitis B viral polymerase. NFAT is a heterodimer of 45 kDa and 90 kDa proteins, the smaller of which is the product of this gene. The encoded protein binds strongly to the 90 kDa protein and stimulates its ability to enhance gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILF2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 5P1K2
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ILF2
Gene Accession No. Q12905, Q7TP98, Q9CXY6
Gene Alias 6230405A16Rik; Ab1-143; ILF2; interleukin enhancer binding factor 2; interleukin enhancer binding factor 2, 45kD; interleukin enhancer binding factor 2, 45kDa; interleukin enhancer-binding factor 2; Interleukin enhancer-binding factor 2 homolog; liver regeneration-related protein LRRG031; MGC8391; Nf45; nuclear factor of activated T-cells 45 kDa; nuclear factor of activated T-cells, 45-kDa; PRO3063; TEG-267; testis expressed gene 267; Tex261; Tex267; Unknown (protein for MGC:134524); wu:fb34h01; zgc:73084; zgc:77276
Gene Symbols ILF2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ILF2 (Q12905).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310612, 3608, 67781
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.