Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541785 Supplier Invitrogen™ Supplier No. PA541785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PTQEELEAVQ NMVSHTERAL KAVSDWIDEQ EKGSSEQAES DNMDVPPEDD Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Rabbit: 85%; Rat: 93%.

UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins. UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility. UBA6 is highly expressed in human monocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ILF3
Gene Accession No. Q12906
Gene Alias CBTF; Double-stranded RNA-binding protein 76; double-stranded RNA-binding protein, 76 kD; DRBF; DRBP76; dsRNA binding protein NFAR-2/MPP4; Ilf3; ilf3 protein; ilf3b; interleukin enhancer binding factor 3; interleukin enhancer binding factor 3, 90kD; interleukin enhancer binding factor 3, 90kDa; interleukin enhancer binding factor 3b; interleukin enhancer-binding factor 3; Interleukin enhancer-binding factor 3 homolog; M phase phosphoprotein 4; MBII-26; MMP4; M-phase phosphoprotein 4; MPHOSPH4; MPP4; NF110; NF110b; NF90; NF90a; NF90b; NFAR; NFAR-1; NFAR2; NF-AT-90; nuclear factor 90; nuclear factor associated with dsRNA; Nuclear factor of activated T-cells 90 kDa; nuclear factor of activated T-cells, 90 kD; TCP110; TCP80; Translational control protein 80; wu:fb37d07; wu:fb94b02; zgc:77030
Gene Symbols ILF3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ILF3 (aa 23-72).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3609
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.