Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILK Antibody (3A9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237448
Description
ILK Monoclonal specifically detects ILK in Human, Mouse, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ILK | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
ILK | |
Purified recombinant fragment of human ILK (AA: 97-244) expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse, Primate, Monkey | |
Ascites |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3A9 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK | |
Mouse | |
51.4 kDa | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
3611 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction