Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060451
Description
ILK Polyclonal specifically detects ILK in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
ILK | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK | |
Rabbit | |
51 kDa | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
3611 | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 2-5 ug/mg lysate | |
Q13418 | |
ILK | |
The immunogen recognized by this antibody maps to a region between residue 402 and 451 of human integrin-linked kinase using the numbering given in entry NP_004508.1 (GeneID 3611). | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Atlantic salmon, Medaka fish, Nile tilapia, Tasmanian devil and Spotted green pufferfish. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction