Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILK Antibody (OTI2H3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271037APC
Description
ILK Monoclonal antibody specifically detects ILK in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ILK | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI2H3 | |
Western Blot | |
59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK | |
Human recombinant protein fragment corresponding to amino acids 1-244 of human ILK (NP_004508) produced in E.coli. | |
0.1 mL | |
Phospho Specific, Protein Kinase, Signal Transduction | |
3611 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction