Learn More
Invitrogen™ ILK Monoclonal Antibody (3A9)

Description
MA5-17099 targets ILK in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human, Mouse, and Non-human primate samples. The MA5-17099 immunogen is purified recombinant fragment of human ILK (amino acids: 97-244) expressed in E. Coli. MA5-17099 detects ILK which has a predicted molecular weight of approximately 51.4kDa.
Specifications
Specifications
| Antigen | ILK |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 3A9 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | ILK |
| Gene Accession No. | O55222, Q13418, Q99J82 |
| Gene Alias | 59 kDa serine/threonine-protein kinase; AA511515; epididymis secretory protein Li 28; ESTM24; HEL-S-28; ILK; ILK1; ILK-1; ILK2; ILK-2; integrin binding protein kinase; Integrin linked ILK; integrin linked kinase; integrin-linked kinase; integrin-linked kinase-2; integrin-linked protein kinase; loc; lost-contact; main squeeze; msq; P59; p59ILK; zgc:65842; zilk |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.