Learn More
ILK Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | ILK |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1962
![]() |
Bethyl Laboratories, Inc
IHC00361T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1961
![]() |
Bethyl Laboratories, Inc
IHC00361 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Zebrafish, Chicken, Sheep, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig,
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
ILK | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
59 kDa serine/threonine-protein kinase, epididymis secretory protein Li 28, HEL-S-28, ILK-1, ILK-2, integrin-linked kinase, integrin-linked kinase-2, P59, p59ILK | |
ILK | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q13418 | |
3611 | |
Between 402 and 451 | |
Primary | |
4° C | |
ILK |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.