Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILKAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106530 Supplier Invitrogen™ Supplier No. PA5106530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ILKAP.

The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILKAP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ILKAP
Gene Accession No. Q8R0F6, Q9H0C8, Q9Z1Z6
Gene Alias 0710007A14Rik; 1600009O09Rik; EC 3.1.3.16; ILK associated serine/threonine phosphatase; Ilkap; ILKAP2; ILKAP3; integrin-linked kinase associated phosphatase; integrin-linked kinase-associated serine/threonine phosphatase; integrin-linked kinase-associated serine/threonine phosphatase 2C; PP2Cdelta; PP2C-DELTA; PPM1O; protein phosphatase 2C; protein phosphatase 2c, delta isozyme; protein phosphatase, Mg2+/Mn2+ dependent 1O
Gene Symbols ILKAP
Host Species Rabbit
Immunogen A synthesized peptide derived from human ILKAP(Accession Q9H0C8), corresponding to amino acid residues L51-S101.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64538, 67444, 80895
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.