Learn More
ILKAP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301479A
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway.Specifications
| ILKAP | |
| Polyclonal | |
| Unconjugated | |
| ILKAP | |
| ILKAP2, ILKAP3, PP2C-DELTA, integrin-linked kinase associated phosphatase, integrin-linked kinase-associated serine/threonine phosphatase 2C, protein phosphatase 2c, delta isozyme | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80895 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9H0C8 | |
| ILKAP | |
| Between 50 and 100 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.