Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILKAP/PP2C delta Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074606JF549
Description
ILKAP/PP2C delta Polyclonal antibody specifically detects ILKAP/PP2C delta in Human samples. It is validated for Western BlotSpecifications
ILKAP/PP2C delta | |
Polyclonal | |
Western Blot | |
DKFZp434J2031, EC 3.1.3.16, FLJ10181, integrin-linked kinase-associated serine/threonine phosphatase, integrin-linked kinase-associated serine/threonine phosphatase 2C, MGC4846, PP2C-DELTA, protein phosphatase 2c, delta isozyme | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human integrin-linked kinase-associated serine/threonine phosphatase 2C using the numbering given in entry NP_110395.1 (GeneID 80895). | |
0.1 mL | |
Signal Transduction | |
80895 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction