Learn More
Description
illustra ExoProStar is our new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits!
The ExoProStar 1-Step PCR and Sequence Reaction Clean-Up reagent provides a simple, single-step method for removing unincorporated primers and nucleotides prior to downstream analysis. illustra Exonuclease I and Alkaline Phosphatase enzymes are combined in a single, stable mix requiring just one pipetting step to set up the clean-up reaction.
illustra ExoProStar 1-Step is based on GE™ founding patents for the clean-up of PCR reactions using Exonuclease 1 and Alkaline Phosphatase. This 1-step version is not available in USA, Canada and Japan. Instead, we offer a version with the enzymes in two separate tubes, the 29002090, and the faster version, 29046321.
Specifications
Specifications
| Format | Tube |
| For Use With (Application) | Enzymatic PCR clean up |
| No. of Reactions | 100 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.