Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva ExoProStar DNA and Sequence Reaction Clean-up
GSA_VA

Catalog No. 45002292 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
20
100
500
2000
5000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-002-292 500
45-002-290 20
45-002-291 100
45-002-293 2000
45-002-294 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-002-292 Supplier Cytiva Supplier No. US78211
Add to Cart
Add to Cart

Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably. illustra™ ExoProStar DNA and Sequence Reaction Clean-up combines enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs.

Enzymes optimized to work together for high efficiency removal of unincorporated primers and dNTPs. Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction.

  • Fast 30-minute protocol
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
  • Complete heat inactivation of the enzymes within 15 minutes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.
For Use With (Application) Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably
Includes One tube of Alkaline Phosphatase and Exonuclease I
No. of Reactions 500
Product Type illustra ExoProStar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.