Learn More
Description
Enzymes optimized to work together for high efficiency removal of unincorporated primers and dNTPs. Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction.
- Fast 30-minute protocol
- Scalable for different reaction sizes
- No loss of PCR product
- Easy to automate
- Complete heat inactivation of the enzymes within 15 minutes
Specifications
Specifications
| Description | Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes. |
| For Use With (Application) | Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably |
| Includes | One tube of Alkaline Phosphatase and Exonuclease I |
| No. of Reactions | 2000 |
| Product Type | illustra ExoProStar |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.