Learn More
Description
Includes
Sample Buffer, reaction buffer, enzyme mix, and control λ DNA
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical DNA of 40 to 50μg, average lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.Thaw components on ice and maintain at 0°C to 4°C during handling. |
| Description | Illustra GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 50 μg yield from nanogram amounts of DNA sample. |
| For Use With (Application) | Genomic DNA Amplification |
| Includes | 10 x 2.25mL Sample Buffer, 10 x 2.25mL Reaction Buffer, 10 x 250μL Enzyme mix, 10 x 20μL Control DNA (lambda) |
| No. of Reactions | 1000 reactions |
| Product Type | illustra GenomiPhi HY DNA Amplification Kit |
| Size | > 10 kb |
| Test Time | 4 h |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
