Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Hot Start Master Mix

Catalog No. 46001001 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-001-001 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-001-001 Supplier Cytiva Supplier No. 25150001
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A ready-to-use 2X mastermix providing robust and reliable performance for demanding PCR applications in which high specificity and high sensitivity are essential to success

  • Convenient, ready-to-use mix saves time and reduces experimental variability
  • For any standard PCR reaction, such as multiplex PCR, and reactions exhibiting nonspecific amplification or primer-dimer formation
  • Primer sequestration effectively reduces any nonspecific priming or primer-dimer formation
  • Recombinant activator protein avoids the risk of contamination from a mammalian source associated with hot start antibodies
  • Avoids the risk of contamination from a mammalian source associated with hot start antibodies
  • Quick hot start activation preserves sample integrity by avoiding extensive precycling heat treatment

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2X Concentrate solution of 20mM Tris HCl (pH 8.6), 100mM KCl, 3mM MgCl2, 0.4mM dNTPs (dATP, dCTP, dGTP, dTTP), 50U/mL Taq DNA Polymerase, hot start activator protein, stabilizers. Store at -20°C
Description Hot Start Master Mix is a 2X premixed formulation that can effectively reduce nonspecific priming and primer-dimer formation during PCR.
Form Solution
Format Solution
Quantity 100 Reactions
For Use With (Application) Standard PCR
No. of Reactions 100
Volume 50μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.