Learn More
Cytiva NAP™ Columns, NAP-25
GSA_VA
Catalog No. 45000149 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
20 Pack
50 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-149 20 Pack
45-000-150 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-149 Supplier Cytiva Supplier No. 17085201
Add to Cart
Edge
Add to Cart
Includes
Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing, and sample elution
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
  • Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
  • Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
  • Useful for small-scale purification, desalting, and buffer exchange
Order Info
Sephadex G-25 DNA grade gel comes in a column buffer consisting of distilled water with 0.15% Kathon™ CG/ICP Biocide.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Rapid and efficient purification of DNA and Oligonucleotides
Includes Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
Product Type NAP-25
Quantity 20 Pack