Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva PuReTaq Ready-To-Go™ PCR Beads
GSA_VA

Catalog No. 46001012 Shop All Cytiva Products
Change view
Click to view available options
Format:
0.2mL Tube with hinged cap
0.2mL Tubes/plate
0.5mL Tube
No. of Reactions:
96
100
480
Volume:
200μL
500μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format No. of Reactions Volume
46-001-012 0.2mL Tubes/plate 480 200μL
46-001-011 0.2mL Tubes/plate 96 200μL
46-001-014 0.2mL Tube with hinged cap 96 200μL
46-001-013 0.5mL Tube 100 500μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 46-001-012 Supplier Cytiva Supplier No. 27955702
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pre-mixed, pre-dispensed reaction beads optimized for performing standard PCR amplifications. Cytiva Lifescience™ illustra™ PuReTaq Ready-To-Go™ PCR Beads are stable at ambient temperature.

  • puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
  • Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
  • Verified for real-time PCR
  • Ready-To-Go means simply add template DNA and primers and cycle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP
Format 0.2mL Tubes/plate
Quantity 1 Kit
For Use With (Application) Standard PCR
No. of Reactions 480
Volume 200μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.