Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Ready-To-Go™ RT-PCR Beads

Optimized for performing either one-step or conventional two-step reverse transcription. Cytiva Lifescience™ Illustra™ Ready-To-Go™ RT-PCR Beads are ambient temperature stable, pre-mixed, and ready for pre-dispensed reactions.

$718.28 - $748.65

Specifications

Quantity 1 Kit
For Use With (Application) RT-PCR
View More Specs

Products 3
Catalog Number Mfr. No. Format No. of Reactions Volume Price Quantity & Availability  
Catalog Number Mfr. No. Format No. of Reactions Volume Price Quantity & Availability  
46-001-005
missing translation for 'viewDocuments'
Cytiva
27925901
0.2mL Tube with hinged cap 96 200μL
Each for $728.04
Only null left
Add to cart
 
46-001-007
missing translation for 'viewDocuments'
Cytiva
27926701
0.2mL Tube 96 200μL
Each for $718.28
Only null left
Add to cart
 
46-001-006
missing translation for 'viewDocuments'
Cytiva
27926601
0.5mL Tube 100 500μL
Each for $748.65
Only null left
Add to cart
 
Description

Description

  • Reagents are optimized for full-length cDNA synthesis (>7.5 kb) and optimal sensitivity from PCR
  • Provided in either thin walled 0.5 ml or 0.2 ml tubes/plates compatible with most thermocyclers
  • Supplied with both pd(N)6 and oligo(dT) primers for flexibility
  • The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage

Specifications

Specifications

1 Kit
RT-PCR
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.