Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Shrimp Alkaline Phosphatase

Catalog No. 45500140 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1000 U
500 U
5000 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-500-140 1000 U
45-500-139 500 U
45-500-138 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-500-140 Supplier Cytiva Supplier No. E70092Z
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10x reaction buffer: 200mM Tris-HCl, pH 8.0, 100mM MgCl2; Dilution buffer: 50mM Tris-HCl, pH 8.0

Catalyzes removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins

  • Bacterial
  • Expressed in E.Coli
  • Inactivated by heating at 75°C for 5 minutes
  • Active in PCR buffers and restriction endonuclease buffers
  • Activity: 1U/μL
  • Purified to homogeneity
  • Free of all contaminating endonucleases, exonucleases and ribonucleases
TRUSTED_SUSTAINABILITY

Specifications

Purity Purified to homogeneity
Source Bacteria
Quantity 1000 U
Product Type Alkaline Phosphatase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.