Learn More
Description
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Specifications
| Format | Liquid |
| Product Type | DNA Polymerization Mix dNTP (20mM each A,C,G,T) |
| Concentration | 20 mM |
| Purity | >99% |
| Quantity | 10 μm |
| Color | Colorless |
| For Use With (Application) | Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.