Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Tissue and Cells genomicPrep Mini Spin Kit

Catalog No. 45001502 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-001-502 50 Preps
45-001-503 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-502 Supplier Cytiva Supplier No. 28904275
Add to Cart
Add to Cart

Includes

Spin columns prepacked with a novel silica-membrane; Lyophilized proteinase K powder; Lysis solutions, wash and elution buffers; 2mL Microcentrifuge collection tubes; Full protocol booklet with a detachable, quick reference protocol card

Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures

To minimize the incidence of DNA shearing during extraction, the illustra tissue and cells genomicPrep Mini Spin Kit includes a lysis solution with proteinase K to gently release genomic DNA from tissues or cells. The kit can be used to extract DNA from diverse tissue types such as liver, kidney, and mouse tails with yields ranging from 0.5 to 1.5μg of genomic DNA per mg of tissue. The kit consistently generates purified genomic DNA with optical density A260/280 readings of about 1.8.

  • Capable of purifying up to 1.5μg/mg of tissue from an input tissue sample range of 5 to 50mg, and up to 40μg of genomic DNA per 5 million cells depending on cell type
  • Total DNA extraction time reduced significantly to 90 minutes (including tissue disruption)
  • Produces high-quality genomic DNA with high molecular weight and less degradation
  • Validated in several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests

TRUSTED_SUSTAINABILITY

Specifications

Description illustra tissue and cells genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures in just 90 min for tissues and 45 min for ce
For Use With (Application) Used for several downstream applications including—restriction enzyme analysis, ligation, cloning, DNA sequencing and PCR
Includes Spin columns prepacked with a novel Silica membrane, Lyophilized Proteinase K powder, Lysis solution, wash and Elution Buffers, microcentrifuge collection tubes, a full protocol booklet and a detachable, quick reference protocol card
No. of Reactions 50 Preps
Product Type illustra tissue and cells genomicPrep Mini Spin Kit
Purity 1.75
Sample Volume 5-50 mg
Test Time 90 min[Animal Tissue], 45 min[Cultured cells]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.