Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iloperidone 98.0+%, TCI America™
SDP

Catalog No. I09261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09261G 1 g
I092625MG 25 mg
I0926100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I09261G Supplier TCI America Supplier No. I09261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133454-47-4
Molecular Formula C24H27FN2O4
Molecular Weight (g/mol) 426.49
MDL Number MFCD00866688
InChI Key XMXHEBAFVSFQEX-UHFFFAOYSA-N
PubChem CID 71360
ChEBI CHEBI:65173
IUPAC Name 1-(4-{3-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]propoxy}-3-methoxyphenyl)ethan-1-one
SMILES COC1=CC(=CC=C1OCCCN1CCC(CC1)C1=NOC2=CC(F)=CC=C12)C(C)=O

Specifications

Melting Point 124°C
Color White
Quantity 1 g
Formula Weight 426.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Iloperidone
RTECSNumber KM5777850
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.