Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILP-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14276
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14276 Supplier Invitrogen™ Supplier No. PA14276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4276 detects the ILP-2 protein in human samples. PA1-4276 has been successfully used in Western blot procedures. PA1-4276 immunogen corresponds to amino acid residues 71-84 of the human ILP-2 protein.

Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family including IAP, XIAP/ILP-1/MIHA, and Livin/KIAP that bind to and inhibits specific proteases. A novel member in the IAP protein family was recently identified and designated ILP-2 for IAP-like protein-2. ILP-2 has high homology to ILP-1, but is encoded by a distinct gene that is solely expressed in testis of tested normal human tissues. ILP-2, unlike ILP-1, has no inhibitory effect on Fas and TNF induced apoptosis, but potently inhibits apoptosis induced by overexpression of Bax or by coexpression of caspase-9 with Apaf-1. ILP-2 interacts with the processed caspase-9. These results suggest that ILP-2 is a novel IAP family member with restricted specificity for caspase-9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILP-2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene BIRC8
Gene Accession No. Q96P09
Gene Alias baculoviral IAP repeat containing 8; baculoviral IAP repeat-containing 8; baculoviral IAP repeat-containing protein 8; BIRC8; hILP2; IAP-like protein 2; ILP2; ILP-2; inhibitor of apoptosis-like protein 2; testis-specific inhibitor of apoptosis
Gene Symbols BIRC8
Host Species Rabbit
Immunogen Synthetic peptide: E(71) E K G H E Y I N N I H L T(84).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112401
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.