Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT-7 (CD85g) Mouse anti-Human, Oligo, Clone: 17G10.2 (also known as 17.2), BD™ AbSeq ™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 940252
This item is not returnable.
View return policy
Description
ILT-7 (CD85g) Monoclonal antibody specifically detects ILT-7 (CD85g) in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
ILT-7 (CD85g) | |
Monoclonal | |
Oligo | |
LILRA4; CD85g; ILT7; ILT-7; Immunoglobulin-like transcript 7 | |
ILT7–FcεRIγ-transfected 2B4 cells | |
RUO | |
Human | |
IgG1 κ |
Single Cell 3' Sequencing | |
Clone 17G10.2 (also known as 17.2) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction