Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT2/CD85j/LILRB1 Mouse anti-Human, Mouse, Rat, Clone: VMP55, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen ILT2/CD85j/LILRB1
Clone VMP55
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug of protein 1ml of lysate
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122472
SDP
View Documents
Novus Biologicals
NBP30749920UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

ILT2/CD85j/LILRB1 Monoclonal specifically detects ILT2/CD85j/LILRB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

ILT2/CD85j/LILRB1
0.2 mg/ml
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Human, Mouse, Rat
CD85 antigen-like family member J, CD85j, CD85j antigen, Ig-like transcript 2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, Monocyte/macrophage immunoglobulin-like receptor 7
Hairy cell leukemia cells
Primary
Store at 4°C.
VMP55
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug of protein 1ml of lysate
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
10859
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.