Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT4/CD85d/LILRB2 Antibody (287219R), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2078RW
Description
ILT4/CD85d/LILRB2 Monoclonal antibody specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOFSpecifications
ILT4/CD85d/LILRB2 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
287219R | |
Western Blot, Flow Cytometry, Neutralization, CyTOF-ready | |
CD85 antigen-like family member D, CD85d antigen, Ig-like transcript 4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4, Gly24-His458, Accession # ACT64556 | |
0.1 mL | |
Immunology | |
10288 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction