Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT5/CD85a/LILRB3 Antibody (222821), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1806Z
Description
ILT5/CD85a/LILRB3 Monoclonal antibody specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
ILT5/CD85a/LILRB3 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11025 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
222821 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
Mouse myeloma cell line NS0-derived recombinant human LILRB3/CD85a/ILT5, Gly24-Glu443, Accession # AAB68668 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction