Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT5/CD85a/LILRB3 Rabbit anti-Mouse, Clone: 58, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | ILT5/CD85a/LILRB3 |
---|---|
Clone | 58 |
Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence |
Applications | Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
ILT5/CD85a/LILRB3 | |
Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
11025 | |
IgG | |
Protein A purified |
58 | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse | |
CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
Recombinant Mouse ILT5/CD85a/LILRB3 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title