Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILT6/CD85e/LILRA3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300150JF646
Description
ILT6/CD85e/LILRA3 Polyclonal antibody specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for ELISASpecifications
ILT6/CD85e/LILRA3 | |
Polyclonal | |
ELISA | |
CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31 | |
Produced in rabbits immunized with purified, recombinant Human ILT6/CD85e/LILRA3 (Accession#: AAH28208.1; Met1-Glu439) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
11026 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction