Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILVBL Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178433AF350
Description
ILVBL Polyclonal specifically detects ILVBL in Human samples. It is validated for Western Blot.Specifications
ILVBL | |
Polyclonal | |
Western Blot | |
acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535 | |
Rabbit | |
Affinity Purified | |
Epigenetics | |
10994.0 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ILVBL | |
A synthetic peptide made to an internal portion of the human ILVBL protein (between residues 350-500) [UniProt A1L0T0] | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction