Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILVBL Monoclonal Antibody (OTI6G9), TrueMAB™, OriGene
SDP

Catalog No. 501674095
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4095 100 μL
50-254-1097 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-4095 Supplier OriGene Supplier No. TA503128
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.

Specifications

Antigen ILVBL
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI6G9
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ILVBL
Gene Accession No. A1L0T0
Gene Alias 209L8, AHAS, HACL1L, ILV2H
Gene Symbols ILVBL
Host Species Mouse
Immunogen Full length human recombit protein of human ILVBL produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10994
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.