Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ILVBL Monoclonal Antibody (OTI8A12)
GREENER_CHOICE

Catalog No. PIMA525585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525585 Supplier Invitrogen™ Supplier No. MA525585
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILVBL Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILVBL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI8A12
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ILVBL
Gene Accession No. A1L0T0
Gene Alias 209L8; 2-hydroxyacyl-CoA lyase 1 like; 2-hydroxyacyl-CoA lyase 2; acetolactate synthase-like protein; 5830463I21; acetolactate synthase homolog; acetolactate synthase-like protein; AHAS; AI415009; HACL1L; ILV2H; ilvB (bacterial acetolactate synthase)-like; ilvB acetolactate synthase like; ILVBL; ilvB-like protein
Gene Symbols ILVBL
Host Species Mouse
Immunogen Full length human recombinant protein of ILVBL produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10994, 362843
Target Species Human, Rat, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.