Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ILVBL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534880
Description
Recommended positive controls: U87-MG, SK-N-SH. Predicted reactivity: Mouse (86%), Rat (88%), Rhesus Monkey (97%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.
Specifications
| ILVBL | |
| Polyclonal | |
| Unconjugated | |
| ILVBL | |
| 209L8; 2-hydroxyacyl-CoA lyase 1 like; 2-hydroxyacyl-CoA lyase 2; acetolactate synthase-like protein; 5830463I21; acetolactate synthase homolog; acetolactate synthase-like protein; AHAS; AI415009; HACL1L; ILV2H; ilvB (bacterial acetolactate synthase)-like; ilvB acetolactate synthase like; ILVBL; ilvB-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10994 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| A1L0T0 | |
| ILVBL | |
| Recombinant protein encompassing a sequence within the center region of human ILVBL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction