Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ILVBL Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305076100UL
Description
ILVBL Polyclonal specifically detects ILVBL in Human, Mouse samples. It is validated for Western Blot.Specifications
ILVBL | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Unconjugated | |
acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 503-632 of human ILVBL (NP_006835.2). | |
100 μL | |
Epigenetics | |
10994 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction