Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva IMAC Sepharose™ 6 Fast Flow Affinity Media

Catalog No. p-4595755 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Particle Size:
45 to 165 μm
90 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Particle Size
45-002-967 100 mL 45 to 165 μm
45-002-966 25 mL 90 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-967 Supplier Cytiva Supplier No. 17092108
Only null left
Add to Cart
Add to Cart

Used for purifying proteins and peptides that have an affinity for metal ions. Cytiva IMAC Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix.

  • Uncharged IMAC resin allows possibility to charge with metal ion of choice for optimized selectivity
  • Used for ions such as histidine-tagged proteins but also untagged recombinant or native proteins
  • High protein binding capacity
  • Coupling chemistry and novel ligand designed to give minimal ligand leakage
  • Available HiTrap™ and HiPrep™ pre-packed formats for easy scale-up and process development

 

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
For Use With (Application) For Purifying Proteins and Peptides That Have an Affinity for Metal Ions
Stability Stable in commonly used aqueous Buffers: 0.01 M HCl, 1 M NaOH, 70% Acetic Acid, 2% SDS, 30% 2-Propanol
Storage Requirements 4°C to 30°C
Product Type IMAC Sepharose 6 Fast Flow Media
Flow Rate 250 to 400 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.