Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Image-iT™ Low Density Lipoprotein Uptake Kit, pHrodo™ Red

Catalog No. I34360
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I34360 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I34360 Supplier Invitrogen™ Supplier No. I34360
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Image-iT Low Density Lipoprotein Uptake Kit - pHrodo Red includes pHrodo Red dye-labeled LDL (also available separately), unlabeled LDL, heparin, BSA, and assay buffer.

The kit was designed to specifically detect binding and uptake of LDL through the LDL receptor internalization pathway, allowing maximum control and flexibility in the experimental design. It can be used with fluorescence microscopy, flow cytometry, cell sorting, high-throughput analysis, and high content analysis.

• pHrodo Red-LDL is detectable with RFP/TRITC filter set (absorption peak: ∼560 nm, emission peak: ∼585 nm)
• pHrodo Red-LDL is multiplexable with green, deep red, violet, and blue emission dyes, such as GFP, DAPI, Hoechst, HCS LipidTOX, and LysoTracker Deep Red
• No-wash protocol is possible, allowing dynamic measurement of LDL uptake and trafficking

pHrodo Red- LDL is dimly fluorescent at neutral pH outside of cells, but fluoresces brightly after endocytosis. This feature allows for a no-wash protocol when using pHrodo Red-LDL, enabling kinetic measurement of LDL uptake, endosomal maturation, and trafficking in cells.

Two controls are included in the kit: unlabeled LDL as an optional pretreatment to occlude cell surface receptors in advance of probing with the labeled construct, and heparin which will chelate the LDL in the solution phase outside of the cells, preventing binding and uptake. When cells are labeled in the absence of these pretreatment controls, a fluorescent signal indicates specific LDL receptor-mediated endocytosis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • 1 x 200 μL pHrodo Red-LDL, 1 mg/mL
  • 1 x 200 μL LDL (unlabeled), 2.5 mg/mL
  • 1 x 50 mL LDL assay buffer, 1 X
  • 1 vial heparin, 5 mg/mL
  • 1 x 150 mg BSA
  • Store in refrigerator (2°C to 8°C) and protect from light.
Product Type Lipoprotein Uptake Kit
Dye Type pH-Sensitive Dyes
Product Line Molecular Probes
Quantity 1 kit
Shipping Condition Wet Ice
Is the LDL in pHrodo Red-LDL acetylated or oxidized?

This product does not contain acetylated or oxidized LDL.

Why is fixation not recommended for cells stained with pHrodo Red-LDL?

pHrodo Red and Green dyes are pH-sensitive dyes. They should only be used on live cells to detect pH of cellular compartments. Upon fixation of the cells, the cell membrane is compromised and you are only measuring the pH of the buffer the samples are held in.

How is the pHrodo Red dye bound to the LDL in pHrodo Red-LDL?

pHrodo Red dye is covalently bound to the LDL.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.