Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Image-iT™ Plant Tissue Hard-Set Mountant

Catalog No. V11331
Encompass_Preferred
Click to view available options
Capacity:
30 mL

Image-iT Plant Tissue Hard-Set Mountant is used for imaging, storage, and archiving of plant tissues optically cleared with Image-iT Plant Tissue Clearing Reagent.

These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue mountant is based on Visikol™ Mount, which has a history of use in fluorescent and colorimetric studies with plants.

• Clears and mounts plant tissue for whole specimen imaging
• Hard-set mountant with refractive index of 1.415

TRUSTED_SUSTAINABILITY

Specifications

Capacity 30 mL
Content And Storage Store at 15°C to 25°C. Do not freeze.
When stored as directed, product is stable for (at least 6 months after receipt.

)
Product Line Image-iT™
Product Type Mountant
Quantity 30 mL
Species Plant
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.