Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Image-iT™ Red Hypoxia Reagent

Catalog No. H10498
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10498 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10498 Supplier Invitrogen™ Supplier No. H10498
  • $712.00
    $683.65 / Each of 1

    Save $28.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Image-iT™ Red Hypoxia Reagent is a novel fluorogenic compound for measuring hypoxia in live cells.

It is non-fluorescent when live cells are in an environment with normal oxygen concentrations and becomes fluorescent when oxygen levels are decreased. Image-iT™ Red Hypoxia Reagent is a real-time oxygen detector, with a fluoregenic response that reverses when the cells return to normal oxygen levels.

Features of Image-iT™ Red Hypoxia Reagent include:

  • Measures hypoxia in live cells by fluorescing in low oxygen environments
  • Real-time oxygen detector, with reversible fluoregenic response
  • Easy to use—just add to cell culture media and image

    Image-iT™ Red Hypoxia Reagent is a fluorogenic compound that becomes fluorescent in environments with low oxygen concentrations, and it is live cell permeable. These properties makes it a highly useful tool for detecting cells and tissue under hypoxic conditions. Image-iT™ Red Hypoxia Reagent is a very sensitive oxygen detector. Unlike pimonidazole adducts that respond only to very low oxygen levels, Image-iT™ Hypoxia Reagent begins to fluoresce when atmospheric oxygen levels are less than 5%. It responds quickly to such environments, and the fluorogenic response reverses if oxygen concentrations improve. These properties make Image-iT™ Red Hypoxia Reagent an ideal tool for detecting hypoxic conditions around tumors, 3D cultures, spheroids, neurons, etc. It can be used to detect tumors in small animals, and its fluorogenic properties have been shown to corresponds with increased Hif 1α expression and translocation in hypoxic environments (1).

    1. Zhang, S., Hosaka, M., Yoshihara, T., Negishi, K., Iida, Y., Tobita, S., & Takeuchi, T. (2010) Phosphorescent light–emitting iridium complexes serve as a hypoxia-sensing probe for tumor imaging in living animals. Cancer Research 70(11), 4490-4498.

  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Provided as 1 mg lyophilized solid. Enough for 3000 assays with 0.1 mL/assay or 600 assays with 0.5 mL/assay
    When stored at ≤-20°C product is stable for (at least 6 months after receipt.
    Protect from light and desiccate.)
    Detection Method Fluorescence
    Product Type Fluorogenic Reagent
    Form Solid
    Product Line Image-iT
    Quantity 1 mg
    Reagent Type Oxygen Sensor
    Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
    Can Image-iT Red Hypoxia Reagent be used in cells expressing fluorescent proteins?

    Yes, as long as the fluorescent protein does not emit in the red range (˜610 nm). For cells that do not express a fluorescent protein, we recommend viewing unstained cells under the TRITC and Texas Red channels to examine autofluorescence. For cells expressing a fluorescent protein, we recommend analyzing unstained cell samples to determine the extent the fluorescent protein emission may overlap in these channels.

    May I include a total cell stain (to stain all cells) along with Image-iT Hypoxia Reagent?

    You may use cell-permeable nuclear counterstains or surface labels, but avoid any general cytoplasmic stains that may interfere with the emission of the hypoxia reagents.

    May I include a dead cell stain along with Image-iT Hypoxia Reagent?

    Yes. We recommend using NucGreen Dead 488 ReadyProbes Reagent (Cat No. R37109) with Image-iT Red Hypoxia Reagent and NucRed Dead 647 ReadyProbes Reagent (Cat No. R37113) with Image-iT Green Hypoxia Reagent.

    Which filter set can I use for optimal excitation and emission with Image-iT Red Hypoxia Reagent?

    The Image-iT Red Hypoxia Reagent has an approximate Ex/Em maxima at 490/610 nm. You may use a 488 nm laser and a Texas Red emission filter. For lamp-based instruments, a YFP longpass filter set may be used (excitation range from 485 to 510 nm and emission from 550 nm up to 620 nm).

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.