Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Imaging System Controller, High Performance

Catalog No. BT1830008 Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
PC:
Z4 G4 Custom Workstation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. PC
BT1830008 Z4 G4 Custom Workstation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BT1830008 Supplier Agilent BioTek Supplier No. 1830008S

This high performance Imaging System Controller is designed for intensive imaging and analysis operations.

  • Microsoft Windows 10 Pro 64-bit Operating System
  • Intel Core i9-7920X Processor (12 core, 2.9 GHz, 16.5 MB cache)
  • Adobe Acrobat Reader DC Pre-installed
  • Ideal for applications such as:
    • Large montages, including single-sample applications
    • Complex image analysis on microplate-based assays
    • High-object count applications like spot counting
    • Higher-throughput imaging application using an Agilent BioTek BioStack microplate stacker or an Agilent BioTek BioSpa automated incubator
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Intensive Imaging and Analysis
Color Black
Includes Keyboard (1), Mouse (1), Front USB 3.1 Port (4), Rear USB 3.1 Port (6), Mini DisplayPort (3), 1 GB LAN Port (2)
Display 24 in. LED Monitor
Operating System Microsoft Windows 10 Pro 64-bit
PC Z4 G4 Custom Workstation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.