Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ IMD 0354
GREENER_CHOICESDP

Catalog No. 261150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-115-0 50 mg
26-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-115-0 Supplier R&D Systems™ Supplier No. 2611/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of IKKβ

Inhibitor of IκB kinase-β (IKKβ) that blocks NF-κB nuclear translocation. Attenuates myocardial ischemia/reperfusion injury by decreasing expression of adhesion molecules ICAM-1 and P-selectin and inhibiting cytokine and chemokine production in cardiomyocytes. Induces G0/G1 cell cycle arrest and apoptosis in HMC-1 and breast cancer cells. Also antagonist at P2X1, P2X4 and P2X7 receptors (IC50 values are 19, 156 and 175 nM respectively). Inhibits platelet aggregation induced by collegen in vitro.

Chemical Identifiers

CAS 978-62-1
Molecular Formula C15H8ClF6NO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 383.67
Purity >99%
Molecular Weight (g/mol) 383.67
Chemical Name or Material IMD 0354
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.