Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iMDK, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-4835938
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Suppressor of MDK expression; inhibits PI 3-K signaling

Suppressor of endogenous midkine (MDK) expression. Inhibits PI 3-K signaling in H441 lung adenocarcinoma cells. Selectively attenuates growth of MDK-expressing cancer cells. Induces apoptosis in H441 cells and suppresses growth of H441 cell xenografts in mice.

Chemical Identifiers

CAS 881970-80-5
Molecular Formula C21H13FN2O2S
Molecular Weight (g/mol) 376.405
InChI Key IWFKQTWYILKFGE-UHFFFAOYSA-N
Synonym imdk, 3-2-4-fluorobenzyl imidazo 2,1-b 1,3 thiazol-6-yl-2h-chromen-2-one, 3-2-4-fluorophenyl methyl imidazo 2,1-b thiazol-6-yl-2h-1-benzopyran-2-one, 3-2-4-fluorophenyl methyl imidazo 2,1-b 1,3 thiazol-6-yl chromen-2-one, 3-2-4-fluorophenyl methyl imidazo 2,1-b 1,3-thiazolin-6-yl chromen-2-one
PubChem CID 15991416
IUPAC Name 3-[2-[(4-fluorophenyl)methyl]imidazo[2,1-b][1,3]thiazol-6-yl]chromen-2-one
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)C3=CN4C=C(SC4=N3)CC5=CC=C(C=C5)F

Specifications

Quantity 10 mg
Formula Weight 376.4
Chemical Name or Material iMDK
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.