Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole, 0.5 M Buffer Solution, pH 7.5, PROMO, Thermo Scientific Chemicals

Catalog No. J63496AEPM
Click to view available options
Quantity:
100 mL
250 mL

Applications

Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

Solubility

Fully miscible.

Notes

Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08 g/mol
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Concentration 0.5 M
pH 7.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material Imidazole
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.