Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2kg
10kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
288-32-4
C3H4N2
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole,glyoxaline,imidazol,iminazole,miazole,1,3-diazole,glyoxalin,imutex,1,3-diaza-2,4-cyclopentadiene,pyrro b monazole
795
CHEBI:16069
1H-imidazole
C1=CN=CN1
288-32-4
C3H4N2
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole,glyoxaline,imidazol,iminazole,miazole,1,3-diazole,glyoxalin,imutex,1,3-diaza-2,4-cyclopentadiene,pyrro b monazole
795
CHEBI:16069
1H-imidazole
C1=CN=CN1
Quantity 5g
Melting Point 86.0°C to 90.0°C
Packaging Glass Bottle
Infrared Spectrum Authentic
Color White to Yellow
Flash Point 145°C
Boiling Point 255.0°C to 256.0°C
Water 0.2% Max. (K.F.)
Vapor Pressure 0.003mbar at 20°C
Assay Percent Range 99%
Beilstein 23, 45
Fieser 01,492; 02,220
Merck Index 15, 4953
Solubility Information Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether
Formula Weight 68.08
Physical Form Flakes
Percent Purity 99%
Chemical Name or Material Imidazole
Show More Show Less

Emergency Overview
Causes burns by all exposure routes. Harmful if swallowed. Possible risk of harm to the unborn child.
NFPA
Health:3 Flammability:1 Instability:0

Signal Word: DANGER!