Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole, 99%

Catalog No. AAA1022136
Click to view available options
Quantity:
100 g
500 g
2500 g

Catalog No. AAA1022136

Supplier: Thermo Scientific Chemicals A1022136

Only null left
Add to Cart

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

Imidazole is used as a buffer in the range of pH 6.2-7.8. It is also an histamine antagonist. It acts as a chelator and forms complexes with various divalent cations. It is used as a corrosion inhibitor on certain transition metals such as copper. Its derivatives, like polybenzimidazole (PBI), act as fire retardants. It finds application in photography and electronics. Imidazole salts are used as ionic liquids and precursors to stable carbenes. Imidazole derivatives like ketoconazole, miconazole and clotrimazole are involved in the treatment of various systemic fungal infections. It is a part of the theophylline molecule, present in tea leaves and coffee beans, which stimulates the central nervous system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole is used as a buffer in the range of pH 6.2-7.8. It is also an histamine antagonist. It acts as a chelator and forms complexes with various divalent cations. It is used as a corrosion inhibitor on certain transition metals such as copper. Its derivatives, like polybenzimidazole (PBI), act as fire retardants. It finds application in photography and electronics. Imidazole salts are used as ionic liquids and precursors to stable carbenes. Imidazole derivatives like ketoconazole, miconazole and clotrimazole are involved in the treatment of various systemic fungal infections. It is a part of the theophylline molecule, present in tea leaves and coffee beans, which stimulates the central nervous system.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, pyridine and methanol. Slightly miscible with benzene and petroleum ether.

Notes
Incompatible with acids, acid anhydrides and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Melting Point 88°C to 92°C
Density 1.03 g/mL
Boiling Point 256°C
Flash Point 145°C (293°F)
Assay Percent Range 99%
Quantity 500 g
Beilstein 103853
Sensitivity Hygroscopic
Solubility Information Miscible with water,ethanol,ether,acetone,chloroform,pyridine and methanol. Slightly miscible with benzene and petroleum ether.
Formula Weight 68.08
Chemical Name or Material Imidazole
Show More Show Less
Hazard Category H302-H314-H335-H360D-H500
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 206-019-2
RTECSNumber NI3325000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.