Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazole, ACS reagent

Catalog No. p-4089452
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Melting Point 86.0°C to 90.0°C
Color White to Yellow
pH 9.5 to 11.0 (at 5% 25°C)
Boiling Point 255.0°C to 256.0°C
Flash Point 145°C
Ignition Residue 0.1% max.
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 4953
Solubility Information Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether
Formula Weight 68.08
Percent Purity ≥99%
Grade ACS Reagent
Physical Form Crystals or Powder
Chemical Name or Material Imidazole, For analysis ACS
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May damage the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
EINECSNumber 206-019-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.