Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Imidazole, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Specifications
Flash Point | 146.00°C |
Packaging | Poly Bottle |
Quantity | 1 kg |
Specific Gravity | 1L = 0.60kg |
Formula Weight | 68.08 |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | Imidazole |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction