Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Imidazole, Reagent, 99%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Imidazole, Reagent is a solid that has no color and is water soluble. It is classified as an diazole and an alkaloid and is an aromatic heterocycle. It belongs to the class of antifungals known as azole and is also used as a corrosion inhibitor on certain metals, notably copper. Imidazole is incorporated into many biological molecules, primarily histidine. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Poly Pail |
Quantity | 1 kg |
UN Number | UN3263 |
Percent Purity | 99% |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction