Learn More
Novus Biologicals™ Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 ligand
Shop All R&D Systems Products

Specifications
Specifications
| Quantity | 5 mg |
| Target | Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 ligand |
| Purity | >98%, by HPLC. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.