Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminodiacetic acid, 98+%

Catalog No. p-7081506
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 142-73-4 | C4H7NO4 | 133.103 g/mol

Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00004280
InChI Key NBZBKCUXIYYUSX-UHFFFAOYSA-N
Synonym iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
PubChem CID 8897
ChEBI CHEBI:24786
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES C(C(=O)O)NCC(=O)O

Specifications

Melting Point ∼243°C (decomposition)
Quantity 100 g
Beilstein 878499
Merck Index 14,4917
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 133.1
Percent Purity ≥98%
Chemical Name or Material Iminodiacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-555-4
RTECSNumber AI2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.