Learn More
Iminodiacetonitrile, 95%
Description
Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.
Solubility
Soluble in water, acetone and hot methanol.
Notes
Incompatible with strong acids and strong oxidizing agents.
Specifications
Specifications
| Melting Point | 75°C to 79°C |
| Quantity | 500 g |
| Beilstein | 773803 |
| Solubility Information | Soluble in water,acetone and hot methanol. |
| Formula Weight | 95.11 |
| Percent Purity | 95% |
| Chemical Name or Material | Iminodiacetonitrile |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| EINECSNumber | 211-058-3 |
| RTECSNumber | AM0650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.